Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Di(4-biphenylyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98.0+%, TCI America™
SDP

Catalog No. D48865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48865G 5 g
D48861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48865G Supplier TCI America Supplier No. D48865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 952431-30-0
Molecular Formula C36H34BNO2
Molecular Weight (g/mol) 523.483
InChI Key PESBFNLGEMHNAM-UHFFFAOYSA-N
Synonym 2-[4-[Di(4-biphenylyl)amino]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-[Di(4-biphenylyl)amino]phenylboronic Acid Pinacol Ester
PubChem CID 67523594
IUPAC Name N,N-bis(4-phenylphenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N(C3=CC=C(C=C3)C4=CC=CC=C4)C5=CC=C(C=C5)C6=CC=CC=C6

Specifications

Color White
Quantity 5 g
Formula Weight 523.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Di(4-biphenylyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.