Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide 98.0+%, TCI America™
SDP

Catalog No. D41525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41525G 5 g
D41521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41525G Supplier TCI America Supplier No. D41525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 34151-49-0 | C24H12N4O4 | 420.384 g/mol

Chemical Identifiers

CAS 34151-49-0
Molecular Formula C24H12N4O4
Molecular Weight (g/mol) 420.384
InChI Key IBRPEOCBRYYINT-UHFFFAOYSA-N
PubChem CID 1381898
SMILES C1=CC2=C3C(=CC=C4C3=C1C(=O)N(C4=O)C5=CC=NC=C5)C(=O)N(C2=O)C6=CC=NC=C6

Specifications

Melting Point 353°C
Quantity 5 g
Formula Weight 420.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.