Learn More
Strem, An Ascensus Company CAS# 1167987-07-6. 500mg. N,N'-Di-t-butyl-2,3-diaminobutane, 98%. MFCD18827623
SDP
Supplier:  Strem, An Ascensus Company 070385500MG
Encompass
CAS# 1167987-07-6. 500mg. N,N'-Di-t-butyl-2,3-diaminobutane, 98%. MFCD18827623. Molecular Weight: 200.36. Molecular Formula: (C4H9)NHCH(CH3)CH(CH3)NH(C4H9). Color/form: colorless, viscous liq. Strem# 07-0385. http://www.strem.com/catalog/v/07-0385/
Catalog No. 50-365-9