Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N'-Di-Tert-Butylcarbodiimide

Catalog No. 2355635G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-556-35G 5 g
23-556-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 23-556-35G Supplier Sigma Aldrich Supplier No. 2355635G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N'sa-Di-tert-butylcarbodiimide can be used:As a reagent for the guanylation of aryl amines catalyzed by lanthanum amides.To prepare dichloroimidazolidine-4,5-dione by reacting with oxalyl chloride, which is a key intermediate for the synthesis of N,N'sa-diamidocarbenes.To prepare the iridium complex of benzamidine named Ir(FMeppy)2(N,N'sa-di-tert-butyl-4-methyl-benzamidine).

Specifications

CAS 691-24-7
Density 0.8 g/mL at 25°C
Boiling Point 48°C to 50°C (12 mmHg, lit.)
Molecular Formula C9H18N2
Refractive Index n20/D 1.428
Linear Formula (CH3)3CN=C=NC(CH3)3
MDL Number MFCD00010231
Quantity 5 g
Molecular Weight (g/mol) 154.25 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.