Learn More
AdipoGen N N-Di-tert-butylthiourea
SDP
Catalog No. 502561120
Encompass_Preferred
Chemical. CAS 4041-95-6. Formula C9H20N2S. MW 188.3. Synthetic Thiourea derivative. Reagent used for synthesis of carbodiimides and isothiocyanate. Used for extraction of noble metals.
Catalog No. 50-256-1120 Supplier AdipoGen Supplier No. CDXD0319G050
Chemical. CAS 4041-95-6. Formula C9H20N2S. MW 188.3. Synthetic Thiourea derivative. Reagent used for synthesis of carbodiimides and isothiocyanate. Used for extraction of noble metals.