Learn More
Medchemexpress LLC HY-114348 100mg Medchemexpress, N,N'-Diacetyl-L-cystine CAS:5545-17-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114348100MG
Encompass_Preferred
Medchemexpress, HY-114348 100mg N,N'-Diacetyl-L-cystine CAS:5545-17-5 N,N'-diacetyl-L-cystine (DiNAC) is the disulphide dimer of N-acetylcysteine with immunomodulating properties. N,N'-diacetyl-L-cystine is a potent, orally active modulator of contact sensitivity/delayed type hypersensitivity reactions in rodents. N,N'-diacetyl-L-cystine also has antiatherosclerotic effects in Watanabe-heritable hyperlipidemic rabbit (WHHL) rabbits. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9332