Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diacetylglycine Anhydride 98.0+%, TCI America™
SDP

Catalog No. D107310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D107310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D107310G Supplier TCI America Supplier No. D107310G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3027-05-2
Molecular Formula C8H10N2O4
Molecular Weight (g/mol) 198.18
MDL Number MFCD00059789
InChI Key CBBKKVPJPRZOCM-UHFFFAOYSA-N
Synonym 1,4-Diacetyl-2,5-diketopiperazine, 1,4-Diacetyl-2,5-dioxopiperazine, 1,4-Diacetyl-2,5-piperazinedione
PubChem CID 540371
IUPAC Name 1,4-diacetylpiperazine-2,5-dione
SMILES CC(=O)N1CC(=O)N(CC1=O)C(=O)C

Specifications

Melting Point 103°C
Quantity 10 g
Formula Weight 198.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diacetylglycine Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.