Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diacetylsulfanilamide 98.0+%, TCI America™
SDP

Catalog No. D177325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D177325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D177325G Supplier TCI America Supplier No. D177325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29591-86-4
Molecular Formula C10H12N2O4S
Molecular Weight (g/mol) 256.276
MDL Number MFCD00008678
InChI Key YTJJPFRVBNFIBN-UHFFFAOYSA-N
PubChem CID 101240
IUPAC Name N-[4-(acetylsulfamoyl)phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC(=O)C

Specifications

Color White
Quantity 25 g
Formula Weight 256.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diacetylsulfanilamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.