Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diallyl-L-tartardiamide, 99%

Catalog No. p-7081977
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1219518 50 g
AAA1219530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1219518 Supplier Thermo Scientific Chemicals Supplier No. A1219518
Only null left
Add to Cart

CAS: 58477-85-3 | C10H16N2O4 | 228.25 g/mol

Cross-linking co-monomer for the polymerization of soluble polyacrylamide gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58477-85-3
Molecular Formula C10H16N2O4
Molecular Weight (g/mol) 228.25
MDL Number MFCD00008640
InChI Key ZRKLEAHGBNDKHM-OCAPTIKFSA-N
Synonym n,n'-diallyltartardiamide, 2r,3s-2,3-dihydroxy-n,n'-bis prop-2-enyl butanediamide, 2r,3s-2,3-dihydroxy-n,n'-bis prop-2-en-1-yl succinamide
PubChem CID 6994946
IUPAC Name (2S,3S)-2,3-dihydroxy-N,N'-bis(prop-2-enyl)butanediamide
SMILES O[C@@H]([C@@H](O)C(=O)NCC=C)C(=O)NCC=C

Specifications

Melting Point 184°C to 186°C
Quantity 50 g
Beilstein 1712934
Optical Rotation +118° (c=3 in methanol)
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material N,N'-Diallyl-L-tartardiamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 261-277-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.