Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-N,N'-Dibenzyl-L-tartaric diamide, 99%, 99% e.e., Thermo Scientific™

Catalog No. AC426980050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426980050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426980050 Supplier Thermo Scientific Chemicals Supplier No. 426980050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88393-56-0
Molecular Formula C18H20N2O4
Molecular Weight (g/mol) 328.37
MDL Number MFCD00064478
InChI Key BBYSAHVLSFBCMN-MNSHXRSINA-N
Synonym 2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide, dibenzyl tartramide, n,n'-dibenzyl-l-tartramide, +-n,n-dibenzyl-l-tartaric diamide, +-n,n'-dibenzyl-l-tartaric diamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide, n,n inverted exclamation marka-dibenzyl-l-tartramide, +-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide
PubChem CID 7271811
IUPAC Name (2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide
SMILES O[C@H]([C@@H](O)C(=O)NCC1=CC=CC=C1)C(=O)NCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 202°C to 206°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Specific Rotation +80° to +86° (20°C, 589nm) (c=1, DMF)
Formula Weight 328.37
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material (+)-N, N'-Dibenzyl-L-tartaric diamide, 99% ee
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.