Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dibenzylaniline 99.0+%, TCI America™
SDP

Catalog No. D0154500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0154500G 500 g
D015425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0154500G Supplier TCI America Supplier No. D0154500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 91-73-6 | Formula: C20H19N | Weight: 273.38 g/mol

Chemical Identifiers

CAS 91-73-6
Molecular Formula C20H19N
Molecular Weight (g/mol) 273.38
MDL Number MFCD00022015
InChI Key ISGXOWLMGOPVPB-UHFFFAOYSA-N
Synonym dibenzylaniline, n-phenyldibenzylamine, aniline, n,n-dibenzyl, benzenemethanamine, n-phenyl-n-phenylmethyl, dibenzylamine, n-phenyl, benzenamine, n,n-bis phenylmethyl, dibenzylaniline, n,n-bis phenylmethyl, dibenzylanilin, bisbenzylphenylamine, aniline, dibenzyl
PubChem CID 66681
IUPAC Name N,N-dibenzylaniline
SMILES C(N(CC1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C
Color White
Boiling Point 180°C
Quantity 500 g
Formula Weight 273.38
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dibenzylaniline
EINECSNumber (3)-0366
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.