Learn More
Medchemexpress LLC N,N'-Dibenzylethylenediamine | 140-28-3 | 99.8% | 240.34 | 5 G
SDP
Catalog No. 5000401207
Encompass_Preferred
N,N'-Dibenzylethylenediamine forms a chelator with metals and interacts with penicillin to form a stable and tasteless salt of penicillin. It is suitable for various research applications.
  • Forms a chelator with metals
  • Interacts with penicillin to form a stable and tasteless salt
  • Suitable as a biochemical assay reagent
  • Functions as a chelator
Catalog No. 50-004-01207 Supplier Medchemexpress LLC Supplier No. HY205125G
N,N'-Dibenzylethylenediamine forms a chelator with metals and interacts with penicillin to form a stable and tasteless salt of penicillin. It is suitable for various research applications.
  • Forms a chelator with metals
  • Interacts with penicillin to form a stable and tasteless salt
  • Suitable as a biochemical assay reagent
  • Functions as a chelator