Learn More
Medchemexpress LLC N,N'-Dibenzylethylenediamine | 140-28-3 | MFCD00004771 | 99.83% | 240.34 | 25 G
SDP
Catalog No. 5000402945
Encompass_Preferred
N,N'-Dibenzylethylenediamine is a compound that functions as a chelator with various metals. It also interacts with penicillin, resulting in the formation of a stable and tasteless salt of penicillin.
  • Forms chelator with metals.
  • Interacts with penicillin to create a stable salt.
  • Produces a tasteless salt of penicillin.
Catalog No. 50-004-02945 Supplier Medchemexpress LLC Supplier No. 25GHY20512
N,N'-Dibenzylethylenediamine is a compound that functions as a chelator with various metals. It also interacts with penicillin, resulting in the formation of a stable and tasteless salt of penicillin.
  • Forms chelator with metals.
  • Interacts with penicillin to create a stable salt.
  • Produces a tasteless salt of penicillin.