Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dibutylethylenediamine 98.0+%, TCI America™
SDP

Catalog No. D151725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D151725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D151725ML Supplier TCI America Supplier No. D151725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3529-09-7
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.32
MDL Number MFCD00042827
InChI Key PWNDYKKNXVKQJO-UHFFFAOYSA-N
Synonym n,n-dibutylethylenediamine, n,n-di-n-butylethylenediamine, 2-dibutylaminoethylamine, 2-aminoethyl dibutylamine, 2-di-n-butylaminoethylamine, 2-di-n-butylamino ethylamine, 1,2-ethanediamine, n,n-dibutyl, 5-norbornene-3-methanol, 2-aminoethyl-dibutyl-amine, 2-dibutylamino ethylamine
PubChem CID 77062
IUPAC Name N',N'-dibutylethane-1,2-diamine
SMILES CCCCN(CCCC)CCN

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 mL
UN Number 2735
Formula Weight 172.32
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dibutylethylenediamine
EINECSNumber (2)-3298
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.