Learn More
Medchemexpress LLC N N-Dicyclohexyl-2-b 100g
SDP
Catalog No. 5000205599
Encompass_Preferred
N N-Dicyclohexyl-2-benzothiazolesulfenamide is a compound with reproductive and developmental toxicity that can affect body weight gain reproductive organ weight and multiple reproductive and developmental indicators in rats
Catalog No. 50-002-05599 Supplier Medchemexpress LLC Supplier No. HYW102684100G
N N-Dicyclohexyl-2-benzothiazolesulfenamide is a compound with reproductive and developmental toxicity that can affect body weight gain reproductive organ weight and multiple reproductive and developmental indicators in rats