Learn More
Medchemexpress LLC N N-Dicyclohexyl-2-b 5g
SDP
Catalog No. 5000205611
Encompass_Preferred
N N-Dicyclohexyl-2-benzothiazolesulfenamide is a compound with reproductive and developmental toxicity that can affect body weight gain reproductive organ weight and multiple reproductive and developmental indicators in rats
Catalog No. 50-002-05611 Supplier Medchemexpress LLC Supplier No. HYW1026845G
N N-Dicyclohexyl-2-benzothiazolesulfenamide is a compound with reproductive and developmental toxicity that can affect body weight gain reproductive organ weight and multiple reproductive and developmental indicators in rats