Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dicyclohexylmethylamine

Catalog No. 2949421KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-494-21KG 1 kg
29-494-2250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-494-21KG Supplier Sigma Aldrich Supplier No. 2949421KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dicyclohexylmethylamine has been employed as:
  • Catalyst during o-phenylation of tertiary alcohol with organobismuth(v) compounds
  • Base during pd-catalyzed 5-endo-trig cyclization of 1-(o-bromophenyl)-2-methylprop-2-en-1-ol

Specifications

CAS 7560-83-0
Density 0.912 g/mL (at 25°C (literature))
Boiling Point 265°C (lit.)
Molecular Formula C13H25N
Linear Formula (C6H11)2 NCH3
Refractive Index n20/D 1.49 (literature)
MDL Number MFCD00014289
Quantity 1 kg
Synonym N-Methyldicyclohexylamine
Molecular Weight (g/mol) 195.34
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.