Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dicyclohexylmethylamine 98.0+%, TCI America™
SDP

Catalog No. D0820100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0820100ML 100 mL
D082025ML 25 mL
D0820500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0820100ML Supplier TCI America Supplier No. D0820100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7560-83-0
Molecular Formula C13H25N
Molecular Weight (g/mol) 195.35
MDL Number MFCD00014289
InChI Key GSCCALZHGUWNJW-UHFFFAOYSA-N
Synonym n,n-dicyclohexylmethylamine, n-methyldicyclohexylamine, cyclohexanamine, n-cyclohexyl-n-methyl, dicyclohexylamine, n-methyl, n-methyl dicyclohexylamine, unii-ri95g6j8ff, n-cyclohexyl-n-methylcyclohexylamine, cy2nme, methyldicyclohexylamine, nn-dicyclohexylmethylamine
PubChem CID 24210
IUPAC Name N-cyclohexyl-N-methylcyclohexanamine
SMILES CN(C1CCCCC1)C2CCCCC2

Specifications

Color Yellow
Boiling Point 150°C
Quantity 100 mL
UN Number 2735
Formula Weight 195.35
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dicyclohexylmethylamine
EINECSNumber (3)-3250
RTECSNumber HY4190000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.