Learn More
Cayman Chemical N,N'-Dicyclohexylurea, Purity: ≥98%, CAS: 2387-23-7, 25 g
SDP
Catalog No. 502887190
Encompass_Preferred
A selective sEH inhibitor with IC50 values of 160 and 90 nM for recombinant human and mouse sEH, respectively; at 10 µM, blocks the hydrolysis of cis- and trans-EETs and completely inhibits 14,(15)-EET-induced PPAR activation
Catalog No. 50-288-7190 Supplier Cayman Chemical Supplier No. 1000497125 G
A selective sEH inhibitor with IC50 values of 160 and 90 nM for recombinant human and mouse sEH, respectively; at 10 µM, blocks the hydrolysis of cis- and trans-EETs and completely inhibits 14,(15)-EET-induced PPAR activation