Learn More
Cayman Chemical N,N'-Dicyclohexylurea, Purity: ≥98%, CAS: 2387-23-7, 10 g
SDP
Catalog No. 502891124
Encompass_Preferred
A selective sEH inhibitor with IC50 values of 160 and 90 nM for recombinant human and mouse sEH, respectively; at 10 µM, blocks the hydrolysis of cis- and trans-EETs and completely inhibits 14,(15)-EET-induced PPAR activation
Catalog No. 50-289-1124 Supplier Cayman Chemical Supplier No. 1000497110 G
A selective sEH inhibitor with IC50 values of 160 and 90 nM for recombinant human and mouse sEH, respectively; at 10 µM, blocks the hydrolysis of cis- and trans-EETs and completely inhibits 14,(15)-EET-induced PPAR activation