Learn More
Apexbio Technology LLC N-N-DICYCLOHEXYLUREA-500MG
SDP
Catalog No. 5000568581
Encompass_Preferred
N N -Dicyclohexylurea (CAS No 2387-23-7) is an organic compound often utilized in biomedical research particularly within the field of enzymology It originates from cyclohexylamine and operates by inhibiting specific enzyme pathways including those involved in protein synthesis regulation The compound has shown notable in vitro activity within the nanomolar to low micromolar range making it suitable for various experimental setups Researchers commonly apply it in cell models to investigate its effects and utilize it in preliminary drug screening processes It is also used in animal studies to evaluate potential therapeutic benefits or toxicological impacts Experimental concentrations typically depend on the specific research design allowing flexibility in achieving desired experimental outcomes
Catalog No. 50-005-68581 Supplier Apexbio Technology LLC Supplier No. C3458500
N N -Dicyclohexylurea (CAS No 2387-23-7) is an organic compound often utilized in biomedical research particularly within the field of enzymology It originates from cyclohexylamine and operates by inhibiting specific enzyme pathways including those involved in protein synthesis regulation The compound has shown notable in vitro activity within the nanomolar to low micromolar range making it suitable for various experimental setups Researchers commonly apply it in cell models to investigate its effects and utilize it in preliminary drug screening processes It is also used in animal studies to evaluate potential therapeutic benefits or toxicological impacts Experimental concentrations typically depend on the specific research design allowing flexibility in achieving desired experimental outcomes