Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dicyclohexylurea, 98%

Catalog No. AC113911000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113911000 100 g Glass bottle
AC113910250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113911000 Supplier Thermo Scientific Chemicals Supplier No. 113911000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2387-23-7 | C13H24N2O | 224.348 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2387-23-7
MDL Number MFCD00003829
InChI Key ADFXKUOMJKEIND-UHFFFAOYSA-N
Synonym n,n'-dicyclohexylurea, dicyclohexylurea, urea, n,n'-dicyclohexyl, unii-zv7823vvim, chembl1458, zv7823vvim, urea,3-dicyclohexyl, urea,n'-dicyclohexyl, n,n inverted exclamation marka-dicyclohexylurea, urea, 1,3-dicyclohexyl
PubChem CID 4277
IUPAC Name 1,3-dicyclohexylurea
SMILES C1CCC(CC1)NC(=O)NC2CCCCC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 231°C to 236°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C13H24N2O
Linear Formula C6H11NHCONHC6H11
Quantity 100 g
Beilstein 12, II, 11
Molecular Weight (g/mol) 224.348
Formula Weight 224.35
Percent Purity ≥97.5% (Total base)
Physical Form Liquid
Chemical Name or Material N, N'-Dicyclohexylurea, 98%
Show More Show Less
EINECSNumber 219-213-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.