Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Didodecylmethylamine 85.0+%, TCI America™
SDP

Catalog No. D3338400G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3338400G 400 g
D333825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3338400G Supplier TCI America Supplier No. D3338400G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2915-90-4
Molecular Formula C25H53N
Molecular Weight (g/mol) 367.706
InChI Key UWHRNIXHZAWBMF-UHFFFAOYSA-N
Synonym N-Methyldidodecylamine
PubChem CID 76205
IUPAC Name N-dodecyl-N-methyldodecan-1-amine
SMILES CCCCCCCCCCCCN(C)CCCCCCCCCCCC

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 204°C
Quantity 400 g
Formula Weight 367.71
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Didodecylmethylamine
EINECSNumber (2)-0176
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.