Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N'-Diethyl-1,3-propanediamine

Catalog No. 13842850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-842-850G 50 g
13-842-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-842-850G Supplier Sigma Aldrich Supplier No. 13842850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,nα-diethyl-1,3-propanediamine is a potential ion-exchange displacer.n,nα-diethyl-1,3-propanediamine was used as displacer in parallel batch screening for ribonuclease a and α-chymotrypsinogen a adsorbed together on high-performance sepharose sp
  • It was used to develop robotic high-throughput displacer screen to identify chemically selective displacers for several protein pairs in cation exchange chromatography

Specifications

CAS 10061-68-4
Density 0.819 g/mL (at 25°C (literature))
Molecular Formula C7H18N2
Refractive Index n20/D 1.4374 (literature)
Linear Formula C2H5NH(CH2)3 NHC2H5
MDL Number MFCD00009036
Quantity 50 g
Synonym 1,3-Bis(ethylamino)propane
Molecular Weight (g/mol) 130.23
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.