Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-2,2,2-trifluoroacetamide, 97%, Thermo Scientific™

Catalog No. AC459330050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459330050 5 g
AC459330250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459330050 Supplier Thermo Scientific Chemicals Supplier No. 459330050

Chemical Identifiers

CAS 360-92-9
Molecular Formula C6H10F3NO
Molecular Weight (g/mol) 169.15
InChI Key CODXZFSZJFCVBE-UHFFFAOYSA-N
Synonym n,n-diethyltrifluoroacetamide, acetamide, n,n-diethyl-2,2,2-trifluoro, acmc-1cmeq, diethyl trifluoroacetamide, n,n-diethyltrifluoro-acetamide, trifluoroacetic acid diethylamide, n,n-diethyl-2,2,2-trifluoroacetamide, n,n-diethyl-2,2,2-tris fluoranyl ethanamide
PubChem CID 225443
IUPAC Name N,N-diethyl-2,2,2-trifluoroacetamide
SMILES CCN(CC)C(=O)C(F)(F)F

Specifications

Density 1.1420g/mL
Boiling Point 65.0°C to 67.0°C (24.0 mmHg)
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula F3CC(=O)N(CH2CH3)2
Quantity 5 g
Specific Gravity 1.142
Solubility Information Solubility in water: immiscible.
Formula Weight 169.15
Percent Purity 97%
Chemical Name or Material N,N-Diethyl-2,2,2-trifluoroacetamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and k

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.