Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-3-aminophenol 98.0+%, TCI America™
SDP

Catalog No. D047025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D047025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D047025G Supplier TCI America Supplier No. D047025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An organic chemical building block primarily used for research and development applications.

Chemical Identifiers

CAS 91-68-9
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00002265
InChI Key WAVOOWVINKGEHS-UHFFFAOYSA-N
Synonym 3-diethylamino phenol, phenol, 3-diethylamino, n,n-diethyl-m-aminophenol, n,n-diethyl-3-aminophenol, m-diethylamino phenol, 3-hydroxy-n,n-diethylaniline, phenol, m-diethylamino, n,n-diethyl-3-hydroxyaniline, unii-bqy98q7yu5, ccris 4615
PubChem CID 7062
IUPAC Name 3-(diethylamino)phenol
SMILES CCN(CC)C1=CC=CC(O)=C1

Specifications

Melting Point 74°C
Boiling Point 280°C
Quantity 25 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material N,N-Diethyl-3-aminophenol
EINECSNumber (3)-0679
RTECSNumber SL0550500
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.