Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N-diethyl-3-(hydroxymethyl)benzamide | 72236-22-7 | MFCD00871280 | 98.6% | C12H17NO2 | 10MG
SDP

Catalog No. 5000259315
Encompass_Preferred

N,N-diethyl-3-(hydroxymethyl)benzamide (ω-hydroxy-DEET) is a research-grade drug metabolite of the insect repellent DEET commonly used in pharmacology, toxicology, and analytical studies. The compound is provided as a characterized, small-quantity sample for in vitro experiments and method development, with supporting analytical data available in the product datasheet.

  • High purity (98.6%).
  • Molecular formula C12H17NO2.
  • Molecular weight 207.27 g/mol.
  • CAS number 72236-22-7.
  • Soluble in DMSO.
  • Packaged in small quantities suitable for assay development.
  • Characterization data and datasheet available for traceability.

Catalog No. 50-002-59315 Supplier Medchemexpress LLC Supplier No. HY13661110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N-diethyl-3-(hydroxymethyl)benzamide (ω-hydroxy-DEET) is a research-grade drug metabolite of the insect repellent DEET commonly used in pharmacology, toxicology, and analytical studies. The compound is provided as a characterized, small-quantity sample for in vitro experiments and method development, with supporting analytical data available in the product datasheet.

  • High purity (98.6%).
  • Molecular formula C12H17NO2.
  • Molecular weight 207.27 g/mol.
  • CAS number 72236-22-7.
  • Soluble in DMSO.
  • Packaged in small quantities suitable for assay development.
  • Characterization data and datasheet available for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.