Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Diethyl-3-methylbenzamide

Catalog No. D100951100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D100951100G 100 g
D100951500G 500 g
D1009512KG 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D100951100G Supplier Sigma Aldrich Supplier No. D100951100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-diethyl-3-methylbenzamide (deet) is generally used as an insect repellant
  • It can undergo reduction in the presence of iron catalyst to synthesize n,n-diethyl-3-methylbenzamine

Specifications

CAS 134-62-3
Density 0.998 g/mL (at 20°C (literature))
Boiling Point 111°C (1 mmHg), 160°C (19 mmHg)
Molecular Formula C12H17NO
Refractive Index n20/D 1.523 (literature)
Linear Formula CH3C6H4CON(C2H5)2
MDL Number MFCD00009046
Quantity 100 g
Synonym N,N-Diethyl-m-toluamide; m-Toluic acid diethylamide; DEET
Molecular Weight (g/mol) 191.27
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.