Learn More
Medchemexpress LLC N,N-diethyl-4-(phenyl(piperidin-4-ylidene)methyl)benzamide hydrochloride | 209808-47-9 | >97.0% | 384.94 g/mol | C23H28N2O.HCl | 100 MG
SDP
Catalog No. 5000213594
Encompass_Preferred
AR-M 1000390 hydrochloride is a non-peptidic, δ-selective opioid receptor agonist (hydrochloride salt) used for research into δ-opioid-mediated analgesia and receptor signaling. It is a brain-penetrant derivative of SNC-80 that shows potent agonist activity with low receptor internalization and diminished acute desensitization, making it useful for in vitro and in vivo pharmacology studies.
  • Potent δ-opioid receptor agonist with EC50 ≈ 7.2 nM.
  • Low receptor internalization compared with typical agonists.
  • Reduces inflammation-induced hyperalgesia in animal models.
  • Brain-penetrant following systemic administration.
  • Hydrochloride salt, molecular weight 384.94 g/mol.
  • Purity typically >97% by HPLC.
Catalog No. 50-002-13594 Supplier Medchemexpress LLC Supplier No. HY101039A100MG
AR-M 1000390 hydrochloride is a non-peptidic, δ-selective opioid receptor agonist (hydrochloride salt) used for research into δ-opioid-mediated analgesia and receptor signaling. It is a brain-penetrant derivative of SNC-80 that shows potent agonist activity with low receptor internalization and diminished acute desensitization, making it useful for in vitro and in vivo pharmacology studies.
  • Potent δ-opioid receptor agonist with EC50 ≈ 7.2 nM.
  • Low receptor internalization compared with typical agonists.
  • Reduces inflammation-induced hyperalgesia in animal models.
  • Brain-penetrant following systemic administration.
  • Hydrochloride salt, molecular weight 384.94 g/mol.
  • Purity typically >97% by HPLC.