Learn More
Medchemexpress LLC N N-Diethyl-N-methyl 100g
SDP
Catalog No. 5000204818
Encompass_Preferred
N N-Diethyl-N-methylethanaminium tetrafluoroborate is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is commonly used as a phase transfer catalyst in organic synthesis facilitating the transfer of reactants between immiscible phases Additionally it is used as an electrolyte in electrochemical devices and as a surfactant in detergent formulations Its unique chemical properties make it an important ingredient in a variety of industrial applications including pharmaceuticals agrochemicals and materials science
Catalog No. 50-002-04818 Supplier Medchemexpress LLC Supplier No. HYW101256100G
N N-Diethyl-N-methylethanaminium tetrafluoroborate is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is commonly used as a phase transfer catalyst in organic synthesis facilitating the transfer of reactants between immiscible phases Additionally it is used as an electrolyte in electrochemical devices and as a surfactant in detergent formulations Its unique chemical properties make it an important ingredient in a variety of industrial applications including pharmaceuticals agrochemicals and materials science