Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-o-toluidine 98.0+%, TCI America™
SDP

Catalog No. D05315ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05315ML 5 mL
D053125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05315ML Supplier TCI America Supplier No. D05315ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 606-46-2
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00059227
InChI Key YQYUUNRAPYPAPC-UHFFFAOYSA-N
Synonym 2-Diethylaminotoluene, N,N-Diethyl-2-methylaniline
PubChem CID 69075
IUPAC Name N,N-diethyl-2-methylaniline
SMILES CCN(CC)C1=CC=CC=C1C

Specifications

Melting Point -60°C
Color Yellow
Boiling Point 210°C
Quantity 5 mL
Formula Weight 163.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-o-toluidine
EINECSNumber (3)-0191
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.