Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-p-phenylenediamine Oxalate (Reagent), Fisher Chemical

Catalog No. O235825 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O2358-25 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O2358-25

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62637-92-7
Molecular Formula C10H16N2 · 0.5 C2H2O4
MDL Number MFCD00070242
Synonym p-Amino-Diethylaniline Oxalate

Specifications

Alpha Vector Diethyl-p-phenylenediamine Oxalate
Melting Point 145°C
Color Brown
Assay Percent Range ≥85 %
Packaging Glass Bottle
Linear Formula [(CH3CH2)2NC6H4NH2]2(COOH)2
Quantity 25 g
Formula Weight 418.55
Percent Purity ≥85%
Grade Reagent
Physical Form Powder or Solid
Chemical Name or Material N, N-Diethyl-p-phenylenediamine Oxalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.