Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N-Diethyl-p-phenylenediamine Sulfate | 6283-63-2 | 99.91% | 262.33 | 1 ML
SDP

Catalog No. 5000451516
Encompass_Preferred

N,N-Diethyl-p-phenylenediamine Sulfate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Appearance is a solid, white to light brown in color.
  • Store at 4°C, sealed, and away from moisture.
  • In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).

Catalog No. 50-004-51516 Supplier Medchemexpress LLC Supplier No. HYW00942510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N-Diethyl-p-phenylenediamine Sulfate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Appearance is a solid, white to light brown in color.
  • Store at 4°C, sealed, and away from moisture.
  • In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.