Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Diethylethylenediamine

Catalog No. 112720100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-272-0100G 100 g
11-272-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-272-0100G Supplier Sigma Aldrich Supplier No. 112720100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N-diethylethylenediamine was used to develop melanin targeted PET and single-photon emission computed tomography (SPECT) imaging agents for melanoma.

Specifications

CAS 100-36-7
Density 0.827 g/mL (at 25°C (literature))
Boiling Point 145°C to 147°C (lit.)
Molecular Formula C6H16N2
Refractive Index n20/D 1.436 (literature)
Linear Formula (C2H5)2 NCH2CH2NH2
MDL Number MFCD00008176
Quantity 100 g
Synonym 2-Diethylaminoethylamine
Molecular Weight (g/mol) 116.2
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.