Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylpropargylamine, 97%

Catalog No. AC455580050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455580050 5 g
AC455580250 25 g
AC455581000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC455580050 Supplier Thermo Scientific Chemicals Supplier No. 455580050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 4079-68-9 | C7H13N | 111.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4079-68-9
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.18
InChI Key JZJXKEWVUBVOEH-UHFFFAOYSA-N
PubChem CID 20010

Specifications

Density 0.8110g/mL
Boiling Point 118°C to 120°C
Flash Point 20°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 0.811
Formula Weight 111.18
Percent Purity 97%
Chemical Name or Material N,N-Diethylpropargylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
EINECSNumber 223-804-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.