Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropyl-1,5-pentanediamine, 97%, Thermo Scientific™

Catalog No. AAH5029206
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5029206 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5029206 Supplier Thermo Scientific Chemicals Supplier No. H5029206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 209803-40-7
Molecular Formula C11H26N2
Linear Formula [(CH3)2CH]2NH(CH2)5NH2
Quantity 5g
MDL Number MFCD00048285
UN Number UN2735
Synonym 5-diisopropylamino amylamine, n,n-diisopropyl-1,5-pentanediamine, 5-aminopentyl diisopropylamine, n1,n1-diisopropylpentane-1,5-diamine, 5-aminopentyl bis methylethyl amine, acmc-1cpdx, n1,n1-dsopropylpentane-1,5-damne, n',n'-di propan-2-yl pentane-1,5-diamine, 1,5-pentanediamine,n1,n1-bis 1-methylethyl
InChI Key CIFBCEXYFFKMEB-UHFFFAOYSA-N
SMILES CC(C)N(CCCCCN)C(C)C
IUPAC Name N',N'-di(propan-2-yl)pentane-1,5-diamine
Molecular Weight (g/mol) 186.343
PubChem CID 3790585
Formula Weight 187.35
Percent Purity 97%
Chemical Name or Material N,N-Diisopropyl-1,5-pentanediamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.