Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diisopropylcarbodiimide, 99%

Catalog No. AAA1929214
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1929214 25 g
AAA1929209 10 g
AAA1929218 50 g
AAA1929230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1929214 Supplier Thermo Scientific Chemicals Supplier No. A1929214
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Alfa Aesar N,N'-Diisopropylcarbodiimide, 99%, Quantity: 25g, Boiling Point: 145 deg.C to 148 deg.C, Molecular Weight: 126.203, Percent Purity: 99%, Beilstein: 878281, CAS: 693-13-0, ChEBI: CHEBI:53092, Density: 0.81, DOT Information: Hazard Class: 6.1; Packaging Group: I, EINECS Number: 211-743-7 | CAS: 693-13-0 | C7H14N2 | 126.203 g/mol

N,N'-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

Solubility
Soluble in chloroform, methylene chloride, acetonitrile, dioxane, dimethylformamide andtetrahydrofuran.

Notes
Moisture sensitive. Store in a cool place Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-13-0
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD00065689
InChI Key BDNKZNFMNDZQMI-UHFFFAOYSA-N
Synonym n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
PubChem CID 12734
ChEBI CHEBI:53092
IUPAC Name N,N'-di(propan-2-yl)methanediimine
SMILES CC(C)N=C=NC(C)C

Specifications

Density 0.81
Boiling Point 145°C to 148°C
Flash Point 33°C (91°F)
Odor Stench
Refractive Index 1.433
Quantity 25 g
UN Number UN2929
Beilstein 878281
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methylene chloride,acetonitrile,dioxane,dimethylformamide andtetrahydrofuran.
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material N,N'-Diisopropylcarbodiimide
Show More Show Less
Hazard Category H226-H315-H317-H318-H330-H334-H335
Hazard Statement GHS H Statement
H330-H334-H318-H226-H315-H317-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 211-743-7
RTECSNumber FF2175000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.