Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diisopropylcarbodiimide, MP Biomedicals™

Catalog No. ICN15775580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15775580 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15775580 Supplier MP Biomedicals, Inc Supplier No. 0215775580

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

CAS 693-13-0
Color Yellow
Density 0.806g/mL
Molecular Formula C7H14N2
Quantity 100 g
Synonym n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
InChI Key BDNKZNFMNDZQMI-UHFFFAOYSA-N
SMILES CC(C)N=C=NC(C)C
IUPAC Name N,N'-di(propan-2-yl)methanediimine
Molecular Weight (g/mol) 126.203
PubChem CID 12734
ChEBI CHEBI:53092
Formula Weight 126.2
Physical Form Liquid
Chemical Name or Material N, N'-Diisopropylcarbodiimide
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.