Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Diisopropylmethylphosphonamidic Chloride

Catalog No. 2625281G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-252-81G 1 g
26-252-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 26-252-81G Supplier Sigma Aldrich Supplier No. 2625281G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used for polymer-supported deoxyoligonucleotide and oligoribonucleotide syntheses.

Specifications

CAS 86030-43-5
Density 1.018 g/mL at 25°C
Boiling Point 34°C to 35°C (0.8 mmHg, lit.)
Molecular Formula C7H17ClNOP
Refractive Index n20/D 1.468
Linear Formula [(CH3)2CH]2NP(OCH3)Cl
MDL Number MFCD00009859
Quantity 1 g
Synonym Chloro(diisopropylamino)methoxyphosphine; Methyl N,N-diisopropylchlorophosphoramidite; Methyl N,N-diisopropylphosphoramidochloridite
Molecular Weight (g/mol) 197.64 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.