Learn More
Medchemexpress LLC N,N-dimethyl-1-(4-(4-methyl-1H-pyrazol-1-yl)pyridin-3-yl)piperidine-4-carboxamide | 1613480-70-8 | 99.6% | C17H23N5O | 10MG
SDP
Catalog No. 5000258271
Encompass_Preferred
Cholesterol 24-hydroxylase-IN-1 is a potent, selective small-molecule inhibitor of cholesterol 24-hydroxylase (CYP46A1) supplied as a high-purity research reagent for biochemical and pharmacological investigations. It is intended for use in assay development, target validation, and mechanistic studies where selective modulation of CH24H activity is required.
  • Potent, selective inhibition of cholesterol 24-hydroxylase (CYP46A1).
  • High purity suitable for sensitive biochemical assays.
  • Molecular formula C17H23N5O and molecular weight 313.40 aid characterization.
  • Supplied as a stable powder with recommended cold storage to preserve integrity.
  • Available in small milligram quantities for research applications.
Catalog No. 50-002-58271 Supplier Medchemexpress LLC Supplier No. HY14430910MG
Cholesterol 24-hydroxylase-IN-1 is a potent, selective small-molecule inhibitor of cholesterol 24-hydroxylase (CYP46A1) supplied as a high-purity research reagent for biochemical and pharmacological investigations. It is intended for use in assay development, target validation, and mechanistic studies where selective modulation of CH24H activity is required.
  • Potent, selective inhibition of cholesterol 24-hydroxylase (CYP46A1).
  • High purity suitable for sensitive biochemical assays.
  • Molecular formula C17H23N5O and molecular weight 313.40 aid characterization.
  • Supplied as a stable powder with recommended cold storage to preserve integrity.
  • Available in small milligram quantities for research applications.