Learn More
Medchemexpress LLC N,N-dimethyl-1-[4-(4-methylpyrazol-1-yl)-3-pyridinyl]piperidine-4-carboxamide | 1613480-70-8 | 99.6% | 313.40 g/mol | C17H23N5O | 5 MG
SDP
Catalog No. 5000221563
Encompass_Preferred
Cholesterol 24-hydroxylase-IN-1 is a potent, selective small-molecule inhibitor of cholesterol 24-hydroxylase (CYP46A1) used in biochemical and pharmacological research. The compound is analytically characterized with a defined molecular formula and high purity, enabling use in enzyme inhibition studies and mechanistic investigations.
  • Potent, selective inhibitor of cholesterol 24-hydroxylase.
  • Suitable for biochemical and pharmacological research applications.
  • High purity (>99.5% by LCMS) supporting analytical use.
  • Molecular weight 313.40 g/mol; formula C17H23N5O.
  • Supplied as a 5 mg research pack.
Catalog No. 50-002-21563 Supplier Medchemexpress LLC Supplier No. HY1443095MG
Cholesterol 24-hydroxylase-IN-1 is a potent, selective small-molecule inhibitor of cholesterol 24-hydroxylase (CYP46A1) used in biochemical and pharmacological research. The compound is analytically characterized with a defined molecular formula and high purity, enabling use in enzyme inhibition studies and mechanistic investigations.
  • Potent, selective inhibitor of cholesterol 24-hydroxylase.
  • Suitable for biochemical and pharmacological research applications.
  • High purity (>99.5% by LCMS) supporting analytical use.
  • Molecular weight 313.40 g/mol; formula C17H23N5O.
  • Supplied as a 5 mg research pack.