Learn More
Medchemexpress LLC N,N-Dimethyl-1-naphthylamine | 86-56-6 | 99.9% | 1 G
SDP
Catalog No. 5000411059
Encompass_Preferred
N,N-Dimethyl-1-naphthylamine is an aromatic amine and a dye. It can be used in the nitrate reduction test.
  • For research use only.
  • Not sold to patients.
Catalog No. 50-004-11059 Supplier Medchemexpress LLC Supplier No. HYD02221G
N,N-Dimethyl-1-naphthylamine is an aromatic amine and a dye. It can be used in the nitrate reduction test.
  • For research use only.
  • Not sold to patients.