Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N-Dimethyl-1-naphthylamine | 86-56-6 | 99.9% | 25 G
SDP

Catalog No. 5000414299
Encompass_Preferred

N,N-Dimethyl-1-naphthylamine is an aromatic amine and a dye, commonly used in nitrate reduction tests. It appears as a colorless to light yellow liquid.

  • Molecular weight of 171.24
  • Chemical formula of C12H13N
  • Density of 1.060±0.06 g/cm3
  • Purity of 99.85%

Catalog No. 50-004-14299 Supplier Medchemexpress LLC Supplier No. 25GHYD0222
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N-Dimethyl-1-naphthylamine is an aromatic amine and a dye, commonly used in nitrate reduction tests. It appears as a colorless to light yellow liquid.

  • Molecular weight of 171.24
  • Chemical formula of C12H13N
  • Density of 1.060±0.06 g/cm3
  • Purity of 99.85%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.