Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-3-nitroaniline 99.0+%, TCI America™
SDP

Catalog No. D075925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D075925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D075925G Supplier TCI America Supplier No. D075925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent used as a chemical building block.

Chemical Identifiers

CAS 619-31-8
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00007236
InChI Key CJDICMLSLYHRPT-UHFFFAOYSA-N
Synonym 3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline
PubChem CID 69269
IUPAC Name N,N-dimethyl-3-nitroaniline
SMILES CN(C)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 60°C
Boiling Point 280°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyl-3-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.