Learn More
Medchemexpress LLC N,N-dimethyl-4-(phenyldiazenyl)aniline-d5 | 1398109-08-4 | 99.0% | 230.32 g·mol⁻¹ | C14H10D5N3 | 10 MG
SDP
Catalog No. 5000299700
Encompass_Preferred
Methyl yellow-d5 is the deuterium-labeled analog of methyl yellow intended as a high-purity reference standard for analytical research. It serves as an internal standard or tracer in quantitative workflows and is provided in small, pre-weighed quantities suitable for laboratory use.
  • Deuterium-labeled tracer for quantitative analysis.
  • Purity 99% for reliable analytical results.
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • Available in small pre-weighed pack sizes for precision dosing.
  • Supplied with datasheet and certificate of analysis for traceability.
Catalog No. 50-002-99700 Supplier Medchemexpress LLC Supplier No. HYW025785S10MG
Methyl yellow-d5 is the deuterium-labeled analog of methyl yellow intended as a high-purity reference standard for analytical research. It serves as an internal standard or tracer in quantitative workflows and is provided in small, pre-weighed quantities suitable for laboratory use.
  • Deuterium-labeled tracer for quantitative analysis.
  • Purity 99% for reliable analytical results.
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • Available in small pre-weighed pack sizes for precision dosing.
  • Supplied with datasheet and certificate of analysis for traceability.