Learn More
Apexbio Technology LLC N,N-Dimethylacetamide 127-19-5 200ml
SDP
Catalog No. 5000090137
Encompass_Preferred
N N-Dimethylacetamide (127-19-5) is an organic solvent and FDA-approved pharmaceutical excipient It modulates cellular signaling pathways to exert anti-inflammatory activity N N-Dimethylacetamide demonstrates this activity principally through inhibition of inflammation-related signaling Based on these pharmacological properties N N-Dimethylacetamide holds research potential in studies of high-fat diet-induced metabolic disturbances and neuroinflammatory processes associated with neurodegenerative disorders such as Alzheimer s disease
Catalog No. 50-000-90137 Supplier Apexbio Technology LLC Supplier No. C6720200
N N-Dimethylacetamide (127-19-5) is an organic solvent and FDA-approved pharmaceutical excipient It modulates cellular signaling pathways to exert anti-inflammatory activity N N-Dimethylacetamide demonstrates this activity principally through inhibition of inflammation-related signaling Based on these pharmacological properties N N-Dimethylacetamide holds research potential in studies of high-fat diet-induced metabolic disturbances and neuroinflammatory processes associated with neurodegenerative disorders such as Alzheimer s disease