Learn More
Medchemexpress LLC N,N-dimethylacetamide | 127-19-5 | MFCD00008686 | 87.12 g/mol | C4H9NO | 500 ML
SDP
Catalog No. 5000210201
Encompass_Preferred
N,N-dimethylacetamide (DMAc) is a polar, aprotic organic solvent used in chemical synthesis, polymer processing, and as a pharmaceutical excipient. It is a clear, colorless to slightly yellow liquid that is miscible with water and many organic solvents. Handle according to the product SDS and use appropriate personal protective equipment.
  • Clear, polar aprotic solvent, miscible with water.
  • Common applications: reaction solvent, polymer processing, pharmaceutical excipient.
  • CAS number 127-19-5; formula C4H9NO; molecular weight 87.12 g/mol.
  • Packaged as a 500 mL bottle.
  • Consult the safety data sheet for storage, handling, and PPE requirements.
Catalog No. 50-002-10201 Supplier Medchemexpress LLC Supplier No. HYW042416500ML
N,N-dimethylacetamide (DMAc) is a polar, aprotic organic solvent used in chemical synthesis, polymer processing, and as a pharmaceutical excipient. It is a clear, colorless to slightly yellow liquid that is miscible with water and many organic solvents. Handle according to the product SDS and use appropriate personal protective equipment.
  • Clear, polar aprotic solvent, miscible with water.
  • Common applications: reaction solvent, polymer processing, pharmaceutical excipient.
  • CAS number 127-19-5; formula C4H9NO; molecular weight 87.12 g/mol.
  • Packaged as a 500 mL bottle.
  • Consult the safety data sheet for storage, handling, and PPE requirements.