Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, ≥99.5% (GC), Solstice

Catalog No. 60048506 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-506 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-506 Supplier Solstice Supplier No. 388401L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Density 0.937g/mL at 25°C
Quantity 1 L
Explosion Limit 1.8% (100°F), 11.5% (160°F)
Flash Point 70°C (158°F)
Ignition Point 914°F
Linear Formula CH3CON(CH3)2
Vapor Pressure 4mmHg (38°C), 2mmHg (25°C)
Beilstein 1737614
Formula Weight 87.12g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Hazard Statement P201-P261-P280-P305 + P351 + P338-P308 + P313
RTECSNumber 204-826-4
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.