Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, 99+%, extra pure

Catalog No. AC181070025
Change view
Click to view available options
Quantity:
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181070025 2.5 L Glass Bottle
AC181070050 5 L Plastic Drum
AC181070250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC181070025 Supplier Thermo Scientific Chemicals Supplier No. 181070025
Only null left
Add to Cart

CAS: 127-19-5 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 0.9370g/mL
Quantity 2.5 L
Flash Point 70°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4370 to 1.4390
Residue after Evaporation 0.01% max.
Specific Gravity 0.937
Assay Percent Range 99+%
Vapor Pressure Vapor Pressure: 1.7mbar at 25°C
Linear Formula CH3CON(CH3)2
Viscosity 1.02 mPa.s (20°C)
Water 0.1% max.
Beilstein 04, 59
Fieser 01,270; 02,144; 04,165
Merck Index 15, 3248
Boiling Point 164.0°C to 166.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
Formula Weight 87.12
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less
Hazard Category Signal Word: WARNING!
Hazard Statement GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement Emergency Overview
Combustible liquid. Harmful in contact with skin. Harmful by inhalation. Irritating to eyes. May cause harm to the unborn child.
NFPA
Health:2 Flammability:2 Instability:0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.