Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, 99%, Reagent Grade, Solstice

Catalog No. 60047930 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-930 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-930 Supplier Solstice Supplier No. D1375102.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

pH 4 (at 200.00g/L 20°C)
Melting Point -20°C
Color Colorless to Yellow
Density 0.937g/mL at 25°C
Quantity 2.5 L
Explosion Limit 11.5% (160°F), 1.8% (100°F)
Flash Point 70°C (158°F)
Ignition Point 914°F
Odor Amine-like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 4mmHg (38°C), 2mmHg (25°C)
Linear Formula CH3CON(CH3)2
Beilstein 1737614
Boiling Point 164°C to 166°C (1013 hPa)
Solubility Information Fully miscible in water
Formula Weight 87.12g/mol
Percent Purity 0.99
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Hazard Statement P201-P261-P280-P305 + P351 + P338-P308 + P313
RTECSNumber 204-826-4
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.